1,2-Dimethyl-1H-imidazole-5-carboxylic acid 95%
Proveedor: Apollo Scientific
OR947693-100MG
OR947693-1G
OR947693-250MG
APOSOR947693-100MGEA
219
EUR
APOSOR947693-100MG
APOSOR947693-1G
APOSOR947693-250MG
1,2-Dimethyl-1H-imidazole-5-carboxylic acid 95%
1,2-Dimethyl-1H-imidazole-5-carboxylic acid
Formula:
C₆H₈N₂O₂ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD16659646 Núm. CAS: 122222-09-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...