[2-(1-Pyrrolidinyl)-3-pyridinyl]methanamine 95%
Proveedor: Apollo Scientific
OR949501-1G
OR949501-250MG
APOSOR949501-1GEA
647
EUR
APOSOR949501-1G
APOSOR949501-250MG
[2-(1-Pyrrolidinyl)-3-pyridinyl]methanamine 95%
[2-(1-Pyrrolidinyl)-3-pyridinyl]methanamine
Formula:
C₁₀H₁₅N₃ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD08690249 Núm. CAS: 859850-79-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...