2-(2-Cyanophenoxy)acetic acid 98%
Proveedor: Apollo Scientific
OR951075-1G
OR951075-25G
OR951075-5G
APOSOR951075-1GEA
21.9
EUR
APOSOR951075-1G
APOSOR951075-25G
APOSOR951075-5G
2-(2-Cyanophenoxy)acetic acid 98%
2-(2-Cyanophenoxy)acetic acid
Formula:
C₉H₇NO₃ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD02257373 Núm. CAS: 6574-95-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...