L-Valine methyl ester hydrochloride 98%

Proveedor: Apollo Scientific

OR951448-100G OR951448-500G
APOSOR951448-100GEA 0 EUR
APOSOR951448-100G APOSOR951448-500G
L-Valine methyl ester hydrochloride 98%
L-Valine methyl ester hydrochloride
Formula: C₆H₁₄ClNO₂
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00012497
Núm. CAS: 6306-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR