3'-Metilpropiofenona 96%
Proveedor: Apollo Scientific
OR951894-25G
APOSOR951894-25GEA
139
EUR
APOSOR951894-25G
3'-Metilpropiofenona 96%
3'-Metilpropiofenona
Formula:
C₁₀H₁₂O MW: 148,2017 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00027014 Núm. CAS: 51772-30-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...