2-Ethoxy-3-trimethylsilanylpyridine 95%
Proveedor: Apollo Scientific
OR952330-1G
OR952330-250MG
APOSOR952330-1GEA
364
EUR
APOSOR952330-1G
APOSOR952330-250MG
2-Ethoxy-3-trimethylsilanylpyridine 95%
2-Ethoxy-3-trimethylsilanylpyridine
Formula:
C₁₀H₁₇NOSi Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD06659011 Núm. CAS: 782479-88-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...