1-Isopropyl-2,5-dimethyl-1H-pyrrole-3-carbaldehyde 95%
Proveedor: Apollo Scientific
OR952532-1G
OR952532-250MG
OR952532-5G
APOSOR952532-1GEA
284
EUR
APOSOR952532-1G
APOSOR952532-250MG
APOSOR952532-5G
1-Isopropyl-2,5-dimethyl-1H-pyrrole-3-carbaldehyde 95%
1-Isopropyl-2,5-dimethyl-1H-pyrrole-3-carbaldehyde
Formula:
C₁₀H₁₅NO Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD06380317 Núm. CAS: 18870-77-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...