Ethyl (4-methylpiperazin-1-yl)acetate 98%
Proveedor: Apollo Scientific
OR953168-10G
OR953168-1G
OR953168-5G
APOSOR953168-10GEA
754
EUR
APOSOR953168-10G
APOSOR953168-1G
APOSOR953168-5G
Ethyl (4-methylpiperazin-1-yl)acetate 98%
Ethyl (4-methylpiperazin-1-yl)acetate
Formula:
C₉H₁₈N₂O₂ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD01755876 Núm. CAS: 28920-67-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...