Dimethyl-N,N-diisopropylphosphoramidite 97%

Proveedor: Apollo Scientific
Atención

Sinónimos: Dimethyl n,n-diisopropylphosphoramidite

OR963457-1G OR963457-5G OR963457-250MG
APOSOR963457-1GEA 120 EUR
APOSOR963457-1G APOSOR963457-5G APOSOR963457-250MG
Dimethyl-N,N-diisopropylphosphoramidite 97%
Dimethyl-N,N-diisopropylphosphoramidite
Formula: C₈H₂₀NO₂P
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00153505
Núm. CAS: 122194-07-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR