3-methyl-5-(trifluoromethyl)benzoyl chloride 97%
Proveedor: Apollo Scientific
PC0467-5G
PC0467-1G
APOSPC0467-5GEA
249
EUR
APOSPC0467-5G
APOSPC0467-1G
3-methyl-5-(trifluoromethyl)benzoyl chloride 97%
3-methyl-5-(trifluoromethyl)benzoyl chloride
Formula:
C₉H₆ClF₃O MW: 222,591 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD1631600 Núm. CAS: 261952-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...