Copper(I) trifluoromethanethiolate 95%
Proveedor: Apollo Scientific
|
Atención
|
PC2096G-1G
PC2096G-250MG
PC2096G-5G
APOSPC2096G-1GEA
313
EUR
APOSPC2096G-1G
APOSPC2096G-250MG
APOSPC2096G-5G
Copper(I) trifluoromethanethiolate 95%
Copper(I) trifluoromethanethiolate
Formula:
CCuF₃S Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00059046 Núm. CAS: 3872-23-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...