Fmoc-2-amino-4,4,4-trifluorobutyric acid
Proveedor: Apollo Scientific
PC27857-100MG
PC27857-1G
PC27857-250MG
APOSPC27857-100MGEA
633
EUR
APOSPC27857-100MG
APOSPC27857-1G
APOSPC27857-250MG
Fmoc-2-amino-4,4,4-trifluorobutyric acid
Fmoc-2-amino-4,4,4-trifluorobutyric acid
Formula:
C₁₉H₁₆F₃NO₄ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD02682447 Núm. CAS: 1219145-37-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...