N-(sec-Butyl)-3-chloro-5-(trifluoromethyl)-2-pyridinamine
Proveedor: Apollo Scientific
PC300583-10G
PC300583-1G
PC300583-5G
APOSPC300583-10GEA
672
EUR
APOSPC300583-10G
APOSPC300583-1G
APOSPC300583-5G
N-(sec-Butyl)-3-chloro-5-(trifluoromethyl)-2-pyridinamine
N-(sec-Butyl)-3-chloro-5-(trifluoromethyl)-2-pyridinamine
Formula:
C₁₀H₁₂ClF₃N₂ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD11118518 Núm. CAS: 1036594-37-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...