Cinacalcet hydrochloride

Proveedor: Apollo Scientific

PC301203-1G PC301203-5G
APOSPC301203-1GEA 61.3 EUR
APOSPC301203-1G APOSPC301203-5G
Cinacalcet hydrochloride
Cinacalcet hydrochloride
Formula: C₂₂H₂₂F₃N·HCl
Número MDL: MFCD08067750
Núm. CAS: 364782-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR