3-Methoxy-4-(trifluoromethyl)benzoic acid 97%
Proveedor: Apollo Scientific
PC302850-5G
PC302850-25G
APOSPC302850-5GEA
84.1
EUR
APOSPC302850-5G
APOSPC302850-25G
3-Methoxy-4-(trifluoromethyl)benzoic acid 97%
3-Methoxy-4-(trifluoromethyl)benzoic acid
Formula:
C₉H₇F₃O₃ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD13185682 Núm. CAS: 276861-63-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...