Hexafluoroglutaryl fluoride 97%
Proveedor: Apollo Scientific
|
Peligro
|
PC4730-25G
PC4730-5G
APOSPC4730-25GEA
893
EUR
APOSPC4730-25G
APOSPC4730-5G
Hexafluoroglutaryl fluoride 97%
Hexafluoroglutaryl fluoride
Formula:
C₅F₈O₂ Pto. Ebullición: 46…47 °C Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00054652 Núm. CAS: 678-78-4 UN: 2922 ADR: 8+6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...