Phenyl trifluoromethyl sulphone 98%
Proveedor: Apollo Scientific
|
Atención
|
Sinónimos:
Phenyl Trifluoromethyl sulphone
PC49074-5G
APOSPC49074-5GEA
119
EUR
APOSPC49074-5G
Phenyl trifluoromethyl sulphone 98%
Phenyl trifluoromethyl sulphone
Formula:
C₇H₅F₃O₂S MW: 210,17 g/mol Densidad: 1,423 g/cm³ (25 °C) Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00159083 Núm. CAS: 426-58-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...