1-(4-Fluorobenzoyl)pyrrolidine 98%
Proveedor: Apollo Scientific
PC901464-100G
PC901464-25G
APOSPC901464-100GEA
538
EUR
APOSPC901464-100G
APOSPC901464-25G
1-(4-Fluorobenzoyl)pyrrolidine 98%
1-(4-Fluorobenzoyl)pyrrolidine
Formula:
C₁₁H₁₂FNO Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00465428 Núm. CAS: 349644-07-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...