(2,6-Difluorophenyl)acetyl chloride
Proveedor: Apollo Scientific
|
Peligro
|
PC9335-1G
PC9335-25G
APOSPC9335-1GEA
91.5
EUR
APOSPC9335-1G
APOSPC9335-25G
(2,6-Difluorophenyl)acetyl chloride
(2,6-Difluorophenyl)acetyl chloride
Formula:
C₈H₅ClF₂O MW: 190,58 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD03424718 Núm. CAS: 116622-90-3 UN: 1760 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...