Tubing, PTFE, b.safe
Proveedor: Bohlender
R750-02 R750-04 R750-06 R750-08
590-0347EA 4.1 EUR
590-0347 590-0348 590-0349 590-0350
Tubing, PTFE, b.safe
Tuberías Tubo de transferencia
Translucent to milky-white appearance.
  • Almost universal chemical resistance
  • Physiologically safe
  • Non-adhesive surface
  • Very good sliding and dielectric characteristics
  • Oxygen value more than 95
  • Resistant to irradiation and weather
  • Can be sterilised in ETO and in autoclaves
Perfect tubing for aggressive and pure liquids or gases.
(1) Bending radius: minimum bending radius in mm at a room temperature of 23 °C
(2) Burst pressure: computed value in bar at a room temperature of 23 °C. It is recommended to restrict the maximum working pressure to 25% of theMore Product Information