Thermomyces lanuginosa lipase
Proveedor: Apollo Scientific
BIIM1002-100G BIIM1002-25G
APOSBIIM1002-100GEA 560 EUR
APOSBIIM1002-100G APOSBIIM1002-25G
Thermomyces lanuginosa lipase
Enzimas
Covalent on IB-150A. Activity: 10000 TBU/g, 5000 PLU/g
  • immobilized on Immobead 150