Single element standards for ICP-OES, selenium
Proveedor: Agilent
5190-8448 5190-8449 5190-8519 5190-8520
HEWL5190-8448EA 95 EUR
HEWL5190-8448 HEWL5190-8449 HEWL5190-8519 HEWL5190-8520
Single element standards for ICP-OES, selenium
Normativas Estándares catiónicos Estándares de selenio
A comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Información de suministro: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Envase: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.