Ceramic Homogenisers
Proveedor: Agilent
5982-9311 5982-9312 5982-9313
HEWL5982-9311EA 52 EUR
HEWL5982-9311 HEWL5982-9312 HEWL5982-9313
Ceramic Homogenisers
Homogenizadores Trituradoras tisulares
QuEChERS ceramic homogenisers increase lab productivity by reducing extraction time while maintaining consistency.
  • Cutting the required extraction time from 60 seconds to as little as 20 seconds – a time savings of 70% per sample
  • Maintaining high, reproducible extractions in a third of the time
  • Minimizing variance between technicians
  • Breaking up salt agglomerates and maintaining a consistent grinding of homogenising material
The same great ceramic homogenisers available in our QuEChERS Kits are also available to buy in bulk, providing excellentMore Product Information