Single element standards for ICP-OES, Samarium
Proveedor: Agilent
5190-8444 5190-8445 5190-8515 5190-8516
HEWL5190-8444EA 0 EUR
HEWL5190-8444 HEWL5190-8445 HEWL5190-8515 HEWL5190-8516
Single element standards for ICP-OES, Samarium
Normativas Estándares catiónicos Estándares de samario
Agilent offers a comprehensive range of single and multi-element standards, tuning solutions, ionization buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of Agilent's standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Información de suministro: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Envase: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.