GC columns, J&W DB-17ht
Proveedor: Agilent
122-1811 122-1831 123-1811 123-1831 123-1861 123-1831E 122-1831LTM
HEWL122-1811EA 543 EUR
HEWL122-1811 HEWL122-1831 HEWL123-1811 HEWL123-1831 HEWL123-1861 HEWL123-1831E HEWL122-1831LTM
GC columns, J&W DB-17ht
Columnas de cromatografía
DB-17ht provides improved resolution for triglycerides and is ideal for confirmational analyses at elevated temperature.
  • (50%-Phenyl)-methylpolysiloxane
  • Mid-polarity
  • Extended upper temperature limit of 365 °C
  • High temperature, polyimide-coated, fused silica tubing
  • Excellent peak shape and faster elution times for high boilers
Agilent JW DB-17ht provides excellent performance criteria for accurate extended temperature range confirmational analyses. The mid-polarity columns are solvent rinsable. With high temperature operation and excellent peak shape delivery, DB-17ht also features faster elution times for high boiling compounds.
Stable and efficient manufacturing utilising high quality components and processes, results in GC columns that feature improved resolution for the effective analysis of triglycerides that remain fully operational over extended temperature limits of 40 to 340/365 °C.