HPLC columns, InfinityLab Poroshell 120 HPH-C8
Proveedor: Agilent
695775-706 695975-506 695975-706 693975-706 699975-506 699975-706 699775-706 693975-506 699770-706 693770-706 693970-506 693970-706 699970-506 699970-706 699975-506K 699975-706K 693975-506K 693975-706K 695770-706 695775-706K 695970-506 695970-706 695975-506K 695975-706K 699775-706K 690770-706 820750-922 820750-929 821725-922 821725-929 823750-922 823750-929 690970-506 690970-706
HEWL695775-706EA 870 EUR
HEWL695775-706 HEWL695975-506 HEWL695975-706 HEWL693975-706 HEWL699975-506 HEWL699975-706 HEWL699775-706 HEWL693975-506 HEWL699770-706 HEWL693770-706 HEWL693970-506 HEWL693970-706 HEWL699970-506 HEWL699970-706 HEWL699975-506K HEWL699975-706K HEWL693975-506K HEWL693975-706K HEWL695770-706 HEWL695775-706K HEWL695970-506 HEWL695970-706 HEWL695975-506K HEWL695975-706K HEWL699775-706K HEWL690770-706 HEWL820750-922 HEWL820750-929 HEWL821725-922 HEWL821725-929 HEWL823750-922 HEWL823750-929 HEWL690970-506 HEWL690970-706
HPLC columns, InfinityLab Poroshell 120 HPH-C8
Columnas de cromatografía
Rugged, high-resolution separations up to five times faster than traditional 5 µm methods.
  • High efficiency columns for improved throughput
  • Excellent lot-to-lot reproducibility gives confidence in separation results
InfinityLab Poroshell 120 columns provide exceptional efficiency and reliability, significantly boosting performance from all instruments, whether need improved HPLC performance, UHPLC performance at lower pressures or the highest UHPLC performance. The three particle diameters 1,9 μm, 2,7 μm, and 4 μm allow to choose the size that best fits separation needs and is most compatible with LC systems, and 12 different chemistries provide a range of selectivities that makes your method development fast and easy.