Single element standards for ICP-OES, Cesium
Proveedor: Agilent
5190-8333 5190-8334 5190-8372 5190-8373
HEWL5190-8333EA 82 EUR
HEWL5190-8333 HEWL5190-8334 HEWL5190-8372 HEWL5190-8373
Single element standards for ICP-OES, Cesium
Normativas Estándares catiónicos Estándares de cesio
Agilent offers a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of Agilent's standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Información de suministro: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Envase: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.