Single element standards for ICP-OES, Iridium
Proveedor: Agilent
5190-8469 5190-8470 5190-9415 5190-8400
HEWL5190-8469EA 119 EUR
HEWL5190-8469 HEWL5190-8470 HEWL5190-9415 HEWL5190-8400
Single element standards for ICP-OES, Iridium
Normativas Estándares catiónicos Estándares de iridio
Agilent offers a comprehensive range of single and multi-element standards, tuning solutions, ionization buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of Agilent's standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Información de suministro: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Envase: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.