Single element standards for ICP-OES, Neodymium
Proveedor: Agilent
5190-8420 5190-8421 5190-8489 5190-8490
HEWL5190-8420EA 0 EUR
HEWL5190-8420 HEWL5190-8421 HEWL5190-8489 HEWL5190-8490
Single element standards for ICP-OES, Neodymium
Normativas Estándares catiónicos Estándares de neodimio
Agilent offers a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of Agilent's standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Información de suministro: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Envase: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.