Single element standards for ICP-OES, Rubidium
Proveedor: Agilent
5190-8440 5190-8441 5190-8511 5190-8512
HEWL5190-8440EA 151 EUR
HEWL5190-8440 HEWL5190-8441 HEWL5190-8511 HEWL5190-8512
Single element standards for ICP-OES, Rubidium
Normativas Estándares catiónicos Estándares de rubidio
Agilent offers a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of Agilent's standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Información de suministro: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Envase: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.