Single element standards for ICP-OES, Vanadium
Proveedor: Agilent
5190-8551 5190-8552 5190-8228 5190-8229
HEWL5190-8551EA 58 EUR
HEWL5190-8551 HEWL5190-8552 HEWL5190-8228 HEWL5190-8229
Single element standards for ICP-OES, Vanadium
Normativas Estándares catiónicos Estándares de vanadio
Agilent offers a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of Agilent's standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Información de suministro: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Envase: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.