Single element standards for ICP-OES, Scandium
Proveedor: Agilent
5190-8446 5190-8447 5190-8517 5190-8518
HEWL5190-8446EA 380 EUR
HEWL5190-8446 HEWL5190-8447 HEWL5190-8517 HEWL5190-8518
Single element standards for ICP-OES, Scandium
Normativas Estándares catiónicos Estándares de escandio
Agilent offers a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
These standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Información de suministro: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Envase: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.