Single element standards for ICP-OES, barium
Proveedor: Agilent
5190-8248 5190-8249 5190-8358 5190-8359
HEWL5190-8248EA 58 EUR
HEWL5190-8248 HEWL5190-8249 HEWL5190-8358 HEWL5190-8359
Single element standards for ICP-OES, barium
Normativas Estándares catiónicos Estándares de bario
Agilent offers a comprehensive range of single and multi-element standards, tuning solutions, ionization buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of Agilent's standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Información de suministro: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Envase: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.