GC columns, J&W DX-1 and DX-4
Proveedor: Agilent
122-6432 122-6462 123-6133 123-6412 123-6432
HEWL122-6432EA 831 EUR
HEWL122-6432 HEWL122-6462 HEWL123-6133 HEWL123-6412 HEWL123-6432
GC columns, J&W DX-1 and DX-4
Columnas de cromatografía
DX-Series of GC columns, the polyethylene glycol (PEG) DX-1 and DX-4 columns, deliver excellent performance over a wide temperature range.
  • DX-1: 90% dimethylpolysiloxane 10% polyethylene glycol
  • DX-4: 15% dimethylpolysiloxane 85% polyethylene glycol
  • Each Agilent J&W GC column is tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency to provide you utmost confidence in qualitative and quantitative results
  • For details about the QC test conditions including the test probe mixture and the resulting chromatogram,