QuEChERS standards
Proveedor: Agilent
5190-0501
5190-0499
5190-0503
HEWL5190-0501EA
353
EUR
HEWL5190-0501
HEWL5190-0499
HEWL5190-0503
QuEChERS standards
Normativas
Estándar para alimentación
Ready-to-use QuEChERS standards are provided for the most commonly used regulatory methods, including AOAC and EN making analysis easier on both GC and LC instruments.
- Save time and inconvenience of making standards
- Available for both GC and LC instruments
- Ready to use for QuEChERS extractions – no dilutions required
In addition to our industry-leading QuEChERS Kits, Agilent makes your analysis easier by providing standards for the most commonly used regulatory methods, including AOAC and EN.