PSM columns, ZORBAX®
Proveedor: Agilent
880957-801 880957-802 880957-805
HEWL880957-801EA 1862 EUR
HEWL880957-801 HEWL880957-802 HEWL880957-805
PSM columns, ZORBAX®
Columnas de cromatografía
Silica based for mechanical stability and no shrinking or swelling with solvent changes. For organic and aqueous solvents.
  • Choices to analyse polymers over the molecular weight range 500 to 100000
  • Alternative to polymeric particles for specialty applications requiring silica's resistance to acids, oxidisers, or reactive species
ZORBAX® PSM columns are manufactured using rugged ZORBAX SIL porous silica microspheres (PSM) and can be used for molecular size separations on most synthetic and natural polymers. Unlike polymeric media, the silica is not swelled by organic or aqueous mobile phases and resists reactive species and strong acids present in certain analyses.