Single element standards for ICP-OES, gadolinium
Proveedor: Agilent
5190-8456 5190-8241 5190-8386 5190-8387
HEWL5190-8456EA 146 EUR
HEWL5190-8456 HEWL5190-8241 HEWL5190-8386 HEWL5190-8387
Single element standards for ICP-OES, gadolinium
Normativas Estándares catiónicos Estándares de gadolinio
Agilent offers a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of Agilent's standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Información de suministro: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Envase: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.