Phenyl-hexyl columns, ZORBAX® Eclipse Plus
Proveedor: Agilent
959758-912 959759-912 959757-912
HEWL959758-912EA 942 EUR
HEWL959758-912 HEWL959759-912 HEWL959757-912
Phenyl-hexyl columns, ZORBAX® Eclipse Plus
Columnas de cromatografía
ZORBAX® Eclipse Plus HPLC columns are the ideal first choice for method development of all samples.
  • High level of performance peak shape, efficiency, resolution, and lifetime with all sample types: acids, bases and neutrals
  • Superior reproducibility with more rigorous QA/QC testing
  • Improved, patented silica manufacturing with start-to-finish product control
  • Three different reversed-phase chemistries are available (Eclipse Plus C18, Eclipse Plus C8 and Eclipse Plus Phenyl-Hexyl)
  • PAH column for high resolution separation of 16 PAHs in EPA 610 - specifically