Captiva collection plates
Proveedor: Agilent
A696001000 A696001000B
HEWLA696001000EA 134 EUR
HEWLA696001000 HEWLA696001000B
Captiva collection plates
Microplacas Placas Deep Well
Captiva 96-well collection plates are designed for use with Captiva filtration plates, SPEC SPE 96-well plates, and Bond Elut 96-well plates.
  • 1 ml format which offers compatibility with further automation or liquid handling
  • Pierceable cover preserves sample integrity
  • Collar vacuum sealed for maximum efficiency and pre-aligned for trouble-free operation
Completely seal the plates, to prevent sample loss due to spillage or evaporation, and sample contamination using Captiva Pierceable 96-well silicone covers. The silicone is specially designed for 96-well autoinjectors, providing easy piercing and removal. Captiva filtration and SPEC 96-well pates can be joined directly onto collection plates using a CaptiVac vacuum collar. The unique design of the Captiva vacuum collar forms a preset, pre-aligned seal between the filtration plate and the collection plate. This seal positions the outlet tips at a specified distance inside each well, to prevent cross contamination of samples.
Precaución: For research use only. Not for use in diagnostic procedures.