Single element standards for ICP-OES, cerium
Proveedor: Agilent
5190-8331 5190-8332 5190-8370 5190-8371
HEWL5190-8331EA 58 EUR
HEWL5190-8331 HEWL5190-8332 HEWL5190-8370 HEWL5190-8371
Single element standards for ICP-OES, cerium
Normativas Estándares catiónicos Estándares de cerio
Agilent offers a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of Agilent's standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Información de suministro: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Envase: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.