GC columns, J&W DB-Petro
Proveedor: Agilent
122-10A6 122-10A6E 128-1056
HEWL122-10A6EA 1532 EUR
HEWL122-10A6 HEWL122-10A6E HEWL128-1056
GC columns, J&W DB-Petro
Columnas de cromatografía
JW DB-Petro is a 100% dimethylpolysiloxane phase, precision-engineered for the analysis of PONA, PIANO and PNA from petroleum processes.
  • These high-resolution DB-Petro columns are bonded, cross-linked, and solvent rinsable, and have been extensively QC tested to the strictest industry standards. DB-Petro also features an operating temperature range of –60 to 325/350 °C.