Single element standards for ICP-OES, copper
Proveedor: Agilent
5190-8348 5190-8349 5190-8378 5190-8379
HEWL5190-8348EA 55 EUR
HEWL5190-8348 HEWL5190-8349 HEWL5190-8378 HEWL5190-8379
Single element standards for ICP-OES, copper
Normativas Estándares catiónicos Estándares de cobre
They offer a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Información de suministro: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Envase: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.