Single element standards for ICP-OES, titanium
Proveedor: Agilent
5190-8545 5190-8546 5190-8224 5190-8225
HEWL5190-8545EA 82 EUR
HEWL5190-8545 HEWL5190-8546 HEWL5190-8224 HEWL5190-8225
Single element standards for ICP-OES, titanium
Normativas Estándares catiónicos Estándares de titanio
They offer a comprehensive range of single and multi-element standards, tuning solutions, ionisation buffers and environmental standards for Atomic Spectroscopy.
  • Gives you confidence in our AAS, ICP-OES or ICP-MS and MP-AES results
  • Accurate, reliable calibrations
All of standards are prepared to exacting specifications that ensure consistency. Every solution is manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
Información de suministro: Supplied with a Material Safety Data Sheet (MSDS) and Certificate of Analysis for complete assurance.
Envase: Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal.