Vannas spring scissors, 3 mm cutting edge
Proveedor: FINE SCIENCE TOOLS
15000-10 15000-00
FSCI15000-10EA 245 EUR
FSCI15000-10 FSCI15000-00
Vannas spring scissors, 3 mm cutting edge
Tijeras
These stainless steel scissors have been designed to maximise visibility under the microscope by eliminating unnecessary bulk in the blades, and they have a long taper which is ideal for cutting vessel walls.