ESD safety shoes, lace-up, Marten XXSports Pro Low
Proveedor: ELTEN
728131/35 728131/36 728131/37 728131/38 728131/39 728131/40 728131/41 728131/42 728131/43 728131/44 728131/45 728131/46 728131/47 728131/48
ELTN728131/35EA 93.9 EUR
ELTN728131/35 ELTN728131/36 ELTN728131/37 ELTN728131/38 ELTN728131/39 ELTN728131/40 ELTN728131/41 ELTN728131/42 ELTN728131/43 ELTN728131/44 ELTN728131/45 ELTN728131/46 ELTN728131/47 ELTN728131/48
ESD safety shoes, lace-up, Marten XXSports Pro Low
Zapatos y botas
Ideal for indoor or outdoor use, in areas with an exposure to moisture, a risk of penetration from pointed and sharp objects, or a risk of electrostatic discharge.Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
  • Full, padded bellows tongue
  • TPU/PU sole, WELLMAXX SPORTS
  • TPU scuff cap
  • Steel toe cap
  • Leather-free
Hydrophobised microfibre/hydrophobised textile material construction with fabric lining for natural ventilation. Full-length inlay sole, SPORTIVE ESD. Non-metallic, penetration-resistant midsole. Lacing system with individual quick locking system. Sole core made of Infinergy® by BASF.
Certificaciones: EN ISO 20345 S3 SRA, form A.