Blackout paper labels
Proveedor: GA
BOP-10C1-2WH BOP-10C3-5WH BOP-127C1-1WH BOP-127C3-2WH BOP-231C1-1WH BOP-231C3-2WH BOP-240C1-2WH BOP-240C3-5WH BOP-242C1-2WH BOP-242C3-5WH BOP-24C1-1WH BOP-24C3-4WH
GAINBOP-10C1-2WHEA 77.2 EUR
GAINBOP-10C1-2WH GAINBOP-10C3-5WH GAINBOP-127C1-1WH GAINBOP-127C3-2WH GAINBOP-231C1-1WH GAINBOP-231C3-2WH GAINBOP-240C1-2WH GAINBOP-240C3-5WH GAINBOP-242C1-2WH GAINBOP-242C3-5WH GAINBOP-24C1-1WH GAINBOP-24C3-4WH
Blackout paper labels
Etiquetas
These cover-up labels are available in roll format, and are compatible with all standard thermal transfer barcode printers.
  • Unique opaque design allows the labels to block out all pre-existing information
  • For general office, laboratory, and warehouse applications
  • Compatible with all major thermal transfer printer brands
Permanent blackout paper labels feature a unique opaque design that allows them to obscure any existing printed information including serial numbers, barcodes, and text. Perfectly suited for general use office, laboratory, and warehouse applications.