Cut resistant gloves, Duracoil 577
Proveedor: Showa
577-10/XXL 577-6/S 577-7/M 577-8/L 577-9/XL
112-6187EA 168 EUR
112-6187 112-6188 112-6189 112-6190 112-6191
Cut resistant gloves, Duracoil 577
Guantes
Innovative Duracoil wrapping technology is used to create this highly cut and abrasion-resistant glove that protects the hand from a range of hazardous applications.
  • Cut, abrasion, and oil-resistant
  • Form fitting, yet breathable and flexible
  • Robust grip, with increased dexterity
  • HPPE reinforced
  • Fully coated nitrile (foam nitrile palm dip)
  • Elasticated wrist cuff
  • Engineered Duracoil liner
  • Machine washable
The nitrile coating is liquid-proof for added protection and comfort, and the foamed nitrile coating provides superior wet and dry grip while remaining breathable and flexible. Ergonomic shaping replicates the hand's shape to reduce hand fatigue and maximize precision.
Ideally suited for use in construction, engineering, manufacturing, mechanical, and oil/gas industries.
Certificaciones: EN 388:2016 (4X42C); Cat. II
Envase: Packaged 10 pair per polybag.