ATEX Special-Duty Air-Operated Double-Diaphragm Pumps
Proveedor: Avantor Fluid Handling
74440-70 74440-72
MFLX74440-70EA 1580 EUR
MFLX74440-70 MFLX74440-72
ATEX Special-Duty Air-Operated Double-Diaphragm Pumps
Bombas Air-Operated Pumps
Unique configurations to meet your demanding needs.
  • High flow rates — up to 106 GPM!
  • Air-powered motor eliminates sparking concerns associated with electrical pumps.
  • Bolted pump design allows easy access to pump head for maintenance and maintains seal after repeated servicing.
ATEX-Approved Nonmetallic Pumps provide 100% groundability with conductive materials of construction and ensuring no static electricity buildup for ignition-free operation. Pumps meet ATEX directives for potentially explosive atmospheres.