Universal microfibre wipes, 14 series
Proveedor: 3M
MFU14GB MFU14GN CC14RT
MMMAMFU14GBEA 227 EUR
MMMAMFU14GB MMMAMFU14GN MMMACC14RT
Universal microfibre wipes, 14 series
Toallitas Paños de limpieza reutilizables Paños para aplicaciones generales
3M™ Universal microfibre wipes, 14 series are versatile and reusable cleaning cloths suitable for use in hospitals, restaurants, offices, and bathrooms.
  • Highly versatile and reusable cloth
  • Ribbon-like fibres penetrate deep into surface irregularities
  • Resists up to 300 wash cycles and machine washable up to 90 °C
Trap and remove dust, dirt, fingerprints, and moisture from a wide variety of surfaces with 3M™ Universal Microfibre Wipes, 14 series. These versatile and reusable cleaning cloths are designed to simultaneously trap and remove soils from a wide variety of surfaces, including delicate surfaces. This results in a long-lasting, streak-free shine. Unlike conventional wipes, the ribbon-like fibres penetrate deep into surface irregularities where dirt, grease, and micro-organisms hide.