Safety glasses, SecureFit™ 500 series
Proveedor: 3M
SF501AF-BLK SF501ASP-BLK SF501SGAF-BLK SF501SGAFBLKFM SF501SGAF-GRN SF501SGAF-RED SF502SGAF-BLK SF505SGAF-DGR SF507SGAF-BLK SF528SGAF-DGR
MMMASF501AF-BLKEA 15.1 EUR
MMMASF501AF-BLK MMMASF501ASP-BLK MMMASF501SGAF-BLK MMMASF501SGAFBLKFM MMMASF501SGAF-GRN MMMASF501SGAF-RED MMMASF502SGAF-BLK MMMASF505SGAF-DGR MMMASF507SGAF-BLK MMMASF528SGAF-DGR
Safety glasses, SecureFit™ 500 series
Gafas
The SecureFit™ 500 series safety glasses feature a stylised lens shape with temples that automatically self-adjust to help provide a secure and comfortable fit.
  • Sleek and compatible safety glasses with adjustable ratchet temples to adjust the lens for facial features and eyeglass position
  • Innovative SecureFit™ pressure diffusion temple technology provides comfortable pressure equalisation
  • Scotchgard™ anti-fog coating provides superior anti-fog and anti-scratch properties (available on selected models)
  • Protection against UV up to 400 nm
  • Wrap-around design improves the security of fit and prevents slipping
  • Flat and flexible temples combine easily with earmuffs and provide solid side protection
  • Easy to clean
Contemporary eye protection designed for comfort and compatibilitys. These safety glasses weigh just 22 g and offer a sleek, stylish look. The pressure diffusion temple technology offers a comfortable fit, with slotted ribs that provide comfortable pressure equalisation, allowing the temples to flex and self-adjust to the size of the wearer’s head. The design of these safety glasses applies pressure to the less sensitive back of the head instead of the temples, achieving a comfortable fit even during rapid head movement. The adjustable ratchet temples offer even greater customisation by allowing users to adjust the lens for facial features and eyeglass position. These glasses combine well with ear muffs.